Learn More
Integrin alpha 5 beta 1 Antibody (BMC5), Novus Biologicals™
Rat Monoclonal Antibody has been used in 2 publications
Brand:  Novus Biologicals NBP2-29788
 View more versions of this product
Product Code. 18767353
  • 6650.00 NOK / 0.10mL
Description
Integrin alpha 5 beta 1 Monoclonal specifically detects Integrin alpha 5 beta 1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, Immunohistochemistry-Paraffin (Negative).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ITGA5
Unpurified
Certificates
Promotions
Promotions
For Research Use Only