missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 5 beta 1 Antibody (BMC5), Novus Biologicals™
Rat Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-29788-0.025ML
This item is not returnable.
View return policy
Description
Integrin alpha 5 beta 1 Monoclonal specifically detects Integrin alpha 5 beta 1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, Immunohistochemistry-Paraffin (Negative).
Specifications
| Integrin alpha 5 beta 1 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with No Preservative | |
| ITGA5 | |
| Mouse embryonal carcinoma P19 cells with myeloma P3X63Ag9.653 cells. | |
| 0.025 mL | |
| Primary | |
| Antibody was raised against mouse integrin VLA-5 (alpha5beta1), a fibronectin receptor. The antibody was prepared by fusion of spleen cells from a Fisher rat immunized with mouse embryonal carcinoma P19 cells with myeloma P3X63Ag9.653 cells. Cell lines tested to express alpha5beta1, include: P19 cells, mouse melanoma B16F1 cells, mouse squamous carcoma SCC cells and thymocytes. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Functional Assay, Immunohistochemistry (Frozen), Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| BMC5 | |
| Western Blot, Flow Cytometry 10ul/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 2 uL, Immunohistochemistry-Frozen 1:10-1:500, Functional 10ul/ml | |
| P08648 | |
| Rat | |
| Unpurified | |
| RUO | |
| 3678 | |
| Mouse | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction