missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ICG 001

Product Code. p-200050503
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18065922

Brand: Tocris Bioscience™ 4505/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits TCF/β-catenin-mediated transcription

ICG 001 is an inhibitor of TCF/β-catenin-mediated transcription. Selectively inhibits β-catenin/CBP interaction; displays no effect on β-catenin/p300 interaction. Exhibits growth inhibitory effects in colon carcinoma cell lines (SW480 and HCT-116 cells); also displays efficacy in Min mouse and nude mouse SW620 xenograft models.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (6S,9aS)-Hexahydro-6-[(4-hydroxyphenyl)methyl]-8-(1-naphthalenylmethyl)-4,7-dioxo-N-(phenylmethyl)-2H-pyrazino[1,2-a]pyrimidine-1(6H)-carboxamide
CAS 780757-88-2
Quantity 10 mg
Target beta-Catenin Inhibitors
Molecular Formula C33H32N4O4
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.