missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ICG 001
Description
ICG 001 is an inhibitor of TCF/β-catenin-mediated transcription. Selectively inhibits β-catenin/CBP interaction; displays no effect on β-catenin/p300 interaction. Exhibits growth inhibitory effects in colon carcinoma cell lines (SW480 and HCT-116 cells); also displays efficacy in Min mouse and nude mouse SW620 xenograft models.
Specifications
Specifications
| Chemical Name or Material | (6S,9aS)-Hexahydro-6-[(4-hydroxyphenyl)methyl]-8-(1-naphthalenylmethyl)-4,7-dioxo-N-(phenylmethyl)-2H-pyrazino[1,2-a]pyrimidine-1(6H)-carboxamide |
| CAS | 780757-88-2 |
| Quantity | 50 mg |
| Target | beta-Catenin Inhibitors |
| Molecular Formula | C33H32N4O4 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?