missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Arylsulfatase A/ARSA Antibody, R&D Systems™

Mouse Monoclonal Antibody
1930.00 NOK - 4225.00 NOK
Specifications
| Antigen | Arylsulfatase A/ARSA |
|---|---|
| Clone | 324802 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18733037
![]() |
R&D Systems
MAB2485-SP |
25 μg |
1930.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18169460
|
R&D Systems
MAB2485 |
100 μg |
4225.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Arylsulfatase A/ARSA Monoclonal specifically detects Arylsulfatase A/ARSA in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Arylsulfatase A/ARSA | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| AAH14210 | |
| 410 | |
| Mouse myeloma cell line NS0-derived recombinant human Arylsulfatase A/ARSA Arg19-Ala507 Accession # AAH14210 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Arylsulfatase A/ARSA in direct ELISAs and Western blots. In direct ELISAs and Western blots, 20-50% cross-reactivity with recombinant mouse ARSA is observed. |
| 324802 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARSA, arylsulfatase A, ASA, Cerebroside 3-sulfatase, cerebroside-sulfatase, EC 3.1.6, EC 3.1.6.8, MLD | |
| ARSA | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title