missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Arylsulfatase A/ARSA Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2485
This item is not returnable.
View return policy
Description
Arylsulfatase A/ARSA Monoclonal specifically detects Arylsulfatase A/ARSA in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Arylsulfatase A/ARSA | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| AAH14210 | |
| ARSA | |
| Mouse myeloma cell line NS0-derived recombinant human Arylsulfatase A/ARSA Arg19-Ala507 Accession # AAH14210 | |
| 100 μg | |
| Primary | |
| Detects human Arylsulfatase A/ARSA in direct ELISAs and Western blots. In direct ELISAs and Western blots, 20-50% cross-reactivity with recombinant mouse ARSA is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 324802 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARSA, arylsulfatase A, ASA, Cerebroside 3-sulfatase, cerebroside-sulfatase, EC 3.1.6, EC 3.1.6.8, MLD | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 410 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction