Learn More
Human Arylsulfatase A/ARSA Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2485-SP
This item is not returnable.
View return policy
Description
Arylsulfatase A/ARSA Monoclonal specifically detects Arylsulfatase A/ARSA in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Arylsulfatase A/ARSA | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| AAH14210 | |
| ARSA | |
| Mouse myeloma cell line NS0-derived recombinant human Arylsulfatase A/ARSA Arg19-Ala507 Accession # AAH14210 | |
| 25 μg | |
| Primary | |
| Detects human Arylsulfatase A/ARSA in direct ELISAs and Western blots. In direct ELISAs and Western blots, 20-50% cross-reactivity with recombinant mouse ARSA is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 324802 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARSA, arylsulfatase A, ASA, Cerebroside 3-sulfatase, cerebroside-sulfatase, EC 3.1.6, EC 3.1.6.8, MLD | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 410 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu