Learn More
Tocris Bioscience™ GYKI 53655 hydrochloride
GREENER_CHOICE
Product Code. 18792524
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18792524 10 mg 10mg
18712794 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18792524 Supplier Tocris Bioscience™ Supplier No. 2555/10
  • ONLINE EXCLUSIVE PRICE
    3580.00 NOK
    3382.05 NOK / 10mg
    Save 197.95 NOK
    5% Off
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
Non-competitive non-NMDA iGluR antagonist
Non-competitive AMPA and kainate receptor antagonist. Analog of GYKI 52466 (Cat. No. 1454). Prolongs the survival time after MgCl2- induced global cerebral ischemia. Exhibits anticonvulsant activity. Also blocks GluK3 homomeric receptors (IC50 = 63 μM) and GluK2b(R)/GluK3 heteroreceptors (IC50 = 32 μM) at high concentrations.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 143692-48-2
Molecular Formula C19H21ClN4O3
Specifications
Quantity 10 mg
Formula Weight 388.85
Purity >99%
Molecular Weight (g/mol) 388.852
Chemical Name or Material GYKI 53655 hydrochloride