missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GYKI 52466 dihydrochloride

Description
GYKI 52466 dihydrochloride is a selective noncompetitive AMPA receptor antagonist (IC50 values are 10-20, ∼ 450 and >50 μM for AMPA-, kainate- and NMDA-induced responses respectively). Skeletal muscle relaxant and orally-active anticonvulsant. Has anti-proliferative effects in transformed cells.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 366.24 |
| Purity | >99% |
| Molecular Weight (g/mol) | 293.326 |
| Chemical Name or Material | GYKI 52466 dihydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?