missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GYKI 52466 dihydrochloride
GREENER_CHOICE

Product Code. 30001256
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
30001256 50 mg 50mg
30001255 10 mg 10mg
2 options
This item is not returnable. View return policy

Product Code. 30001256

Brand: Tocris Bioscience™ 1454/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective non-competitive AMPA antagonist

GYKI 52466 dihydrochloride is a selective noncompetitive AMPA receptor antagonist (IC50 values are 10-20, ∼ 450 and >50 μM for AMPA-, kainate- and NMDA-induced responses respectively). Skeletal muscle relaxant and orally-active anticonvulsant. Has anti-proliferative effects in transformed cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2319722-40-0
Molecular Formula C17H15N3O2

Specifications

Quantity 50 mg
Formula Weight 366.24
Purity >99%
Molecular Weight (g/mol) 293.326
Chemical Name or Material GYKI 52466 dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.