Learn More
Tocris Bioscience™ GW 788388
Product Code. 18085312
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
18085312 50 mg 50mg
18004322 10 mg 10mg
2 options
Product Code. 18085312
Brand: Tocris Bioscience™ 3264/50
Potent and selective inhibitor of TGF-βRI
Potent inhibitor of transforming growth factor-β type I receptor (ALK5) (IC50 values are 18 and 93 nM for ALK5 binding and for TGF-β cellular assay respectively). Inhibits esophageal squamous cell carcinoma (ESCC)-induced neoangiogenesis. Orally active.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 452342-67-5
Molecular Formula C25H23N5O2
Specifications
Quantity 50 mg
Formula Weight 425.48
Purity >98%
Molecular Weight (g/mol) 425.49
Chemical Name or Material GW 788388