Learn More
Gr-1 (Ly-6G/Ly-6C) Rat, anti-Mouse, FITC, Clone: RB6-8C5, STEMCELL Technologies
Rat Monoclonal Antibody
Brand: Stemcell Technologies 60028FI
This item is not returnable.
View return policy
Description
- The RB6-8C5 antibody reacts with the structurally related murine Ly-6G (21 - 25 kDa) and Ly-6C (14 - 16 kDa) GPI-anchored proteins, which together comprise the granulocyte receptor-1 antigen (Gr-1)
- Gr-1 is expressed on monocytes, neutrophils, subsets of macrophages, plasmacytoid dendritic cells, and T cells
- Monocytes in the bone marrow transiently express Gr-1 during development and the expression level is strongly correlated with granulocyte differentiation and maturation
- In the periphery Gr-1 is found predominantly on neutrophils and is a useful marker for these cells
- Whereas the RB6-8C5 binds to both Ly-6G and Ly-6C, another commonly used anti-Gr-1 antibody, clone 1A8, binds specifically to Ly-6G
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Neutrophil Enrichment Kit (Catalog #19762)
Specifications
| Gr-1 (Ly-6G/Ly-6C) | |
| Monoclonal | |
| FITC | |
| 500 ÎĽg | |
| Mouse | |
| Granulocytes and Subsets, Monocytes, Myeloid Cells |
| Immunoprecipitation, Western Blot, Flow Cytometry, Immunocytochemistry | |
| RB6-8C5 | |
| Rat | |
| Primary | |
| IgG2b Îş |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Gr-1 (Ly-6G/Ly-6C) Rat, anti-Mouse, FITC, Clone: RB6-8C5, STEMCELL Technologies