Learn More
Tocris Bioscience™ Go 6976
Product Code. 18094722
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
Product Code. Quantity unitSize
18094722 500 μg 500µg
1 options
Product Code. 18094722
Brand: Tocris Bioscience™ 2253/500U
Potent protein kinase C inhibitor; selective for α and β isozymes
Go 6976 is a potent protein kinase C (PKC) inhibitor (IC50 = 7.9 nM). Discriminates between Ca2+-dependent and -independent isoforms of PKC in vitro; selectively inhibits PKCα and PKCβ1 (IC50 values are 2.3 and 6.2 nM respectively). Does not inhibit the activity of PKCδ, -ε, or -ζ (IC50 > 3μM). Also inhibits TrkA, TrkB, JAK2 and JAK3 tyrosine kinases (IC50 values are 5, 30, 130 and 370 nM respectively), PKCμ (PKD1) and the mitotic spindle checkpoint.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5,6,7,13-Tetrahydro-13-methyl-5-oxo-12H-indolo[2,3-a]pyrrolo[3,4-c]carbazole-12-propanenitrile
CAS 136194-77-9
Quantity 500 μg
Target Protein Kinase C Inhibitors
Molecular Formula C24H18N4O
Purity 95%