Learn More
Tocris Bioscience™ GN 44028
Description
GN 44028 is a potent HIF-1α inhibitor (IC50 = 14 nM). Inhibits hypoxia-induced HIF-1α transcriptional activity without suppressing HIF-1α mRNA expression, HIF-1α protein accumulation, or HIF-1α/HIF-1β heterodimerization. Cytotoxic to a range of cancer cell types in vitro.
Specifications
Specifications
| Chemical Name or Material | N-(2,3-Dihydro-1,4-benzodioxin-6-yl)-1,4-dihydroindeno[1,2-c]pyrazol-3-amine |
| CAS | 1421448-26-1 |
| Quantity | 10 mg |
| Target | HIF Inhibitors |
| Molecular Formula | C18N15N3O2 |
| Formula Weight | 305.33 |
| Purity | >98% |