Learn More
WTW™ Glass Overflow Measuring Flask
Replacement sample bottles and other glassware as well as useful accessories for the expansion of application of measuring systems for different parameters.WTW offers a large assortment of glass ware and other products to replace, supplement and to extend your applications.
539.00 NOK - 810.00 NOK
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
10411602
|
WTW™
209030 |
22.7 mL |
539.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
10327771
|
WTW™
209090 |
432 mL |
810.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
10091931
|
WTW™
209080 |
365 mL |
774.00 NOK
Each |
In Stock
Log in to see stock. |
||||||
10114261
|
WTW™
209070 |
250 mL |
662.00 NOK
Each |
In Stock
Log in to see stock. |
Hai giĂ un account? Accedi per visualizzare il prezzo e la disponibilitĂ . Sei nuovo da queste parti? Registrati qui! |
|||||
10776642
|
WTW™
209040 |
43.5 mL |
539.00 NOK
Each |
In Stock
Log in to see stock. |
Hai giĂ un account? Accedi per visualizzare il prezzo e la disponibilitĂ . Sei nuovo da queste parti? Registrati qui! |
|||||
Descrizione
Specifica
BOD | |
Glass |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.