missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Forskolin
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
Description
- Forskolin is a cell-permeable diterpene that directly activates adenylyl cyclase (IC50 = 41 nM), the enzyme that produces cyclic adenosine monophosphate (cAMP), which as a result raises cAMP levels in the cell
- cAMP is an important second messenger involved in many signal transduction pathways, including activation of protein kinase A (PKA; Awad et al.; Robbins et al.)
Specifications
Specifications
| CAS | 66575-29-9 |
| Synonym | Coleonol; HL 362; L 75-1362B; NSC 357088; NSC 375489 |
| Quantity | 1 mg |
| Molecular Formula | C22H34O7 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction