Learn More
Tocris Bioscience™ Flurofamide
Description
Flurofamide is a urease inhibitor.
Specifications
Specifications
| CAS | 70788-28-2 |
| Quantity | 50 mg |
| Molecular Formula | C7H9FN3O2P |
| Formula Weight | 217.14 |
| Purity | >98% |
| Molecular Weight (g/mol) | 217.14 |
| Chemical Name or Material | Flurofamide |