Learn More
Fisherbrand™ Covers for Fisherbrand™ Ultrasonic Cleaners
Product Code. 11716208 Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
Ultrasonic Cleaner, 0.8 L
Ultrasonic Cleaner, 1.75 L
Ultrasonic Cleaner, 18 L
Ultrasonic Cleaner, 2.75 and 4.25 L
Ultrasonic Cleaner, 28 L
Ultrasonic Cleaner, 45 L
Ultrasonic Cleaner, 5.75 L
Ultrasonic Cleaner, 6.9 and 9.4 L
Ultrasonic Cleaner, 9.5 and 12.75 L
Ultrasonic Cleaner, 90 L
Type:
Lid
Unit Size:
Each
Product Code. 11716208
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A practical cover to complete your S-Line Ultrasonic Cleaning Unit. Fisherbrand™ Covers for S-Line Ultrasonic Cleaning Units are manufactured from durable blue plastic.
  • Easy to clean
  • Clean-cut, smooth surfaces
  • Meets high user demands for hygiene
Specifications
Type Lid
For Use With (Equipment) Ultrasonic Cleaner, 90 L
Material Stainless Steel