Learn More
Fisherbrand™ Covers for Fisherbrand™ Ultrasonic Cleaners
Product Code. 10798484 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Equipment):
Ultrasonic Cleaner, 0.8 L
Ultrasonic Cleaner, 1.75 L
Ultrasonic Cleaner, 18 L
Ultrasonic Cleaner, 2.75 and 4.25 L
Ultrasonic Cleaner, 28 L
Ultrasonic Cleaner, 45 L
Ultrasonic Cleaner, 5.75 L
Ultrasonic Cleaner, 6.9 and 9.4 L
Ultrasonic Cleaner, 9.5 and 12.75 L
Ultrasonic Cleaner, 90 L
Type:
Lid
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Type unitSize
10798484 Ultrasonic Cleaner, 18 L Lid Each
10060892 Ultrasonic Cleaner, 0.8 L Lid Each
10235232 Ultrasonic Cleaner, 1.75 L Lid Each
10560243 Ultrasonic Cleaner, 2.75 and 4.25 L Lid Each
10798294 Ultrasonic Cleaner, 5.75 L Lid Each
11846244 Ultrasonic Cleaner, 6.9 and 9.4 L Lid Each
10487912 Ultrasonic Cleaner, 9.5 and 12.75 L Lid Each
10345962 Ultrasonic Cleaner, 28 L Lid Each
11706208 Ultrasonic Cleaner, 45 L Lid Each
11716208 Ultrasonic Cleaner, 90 L Lid Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10798484
A practical cover to complete your S-Line Ultrasonic Cleaning Unit. Fisherbrand™ Covers for S-Line Ultrasonic Cleaning Units are manufactured from durable blue plastic.
  • Easy-to-open and close cover
  • Muffles noise level
  • Designed to be used as drip basin when flipped over
  • Chemical-resistant plastic
  • Bring your ultrasonic cleaner up to temperature more quickly
  • Contain and prevent spills and leakage
Specifications
Type Lid
For Use With (Equipment) Ultrasonic Cleaner, 18 L
For Use With (Application) Medical Laboratories and in Industrial Production Processes
Material Plastic