Learn More
Fisherbrand™ Stainless Steel Covers for Ultrasonic Units
Product Code. 10248951 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (English):
0.75 gal.
1 gal.
Capacity (Metric):
2.83 L
3.78 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) Capacity (Metric) Unit Size
10248951 1 gal. 3.78 L Each
10101081 0.75 gal. 2.83 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10248951
  • ONLINE EXCLUSIVE PRICE
    482.00 NOK
    305.36 NOK / Pack of 1
    Save 176.64 NOK
    36% Off
Estimated Shipment: 22-01-2027
Add to basket
Add to basket
For use with Fisherbrand 112xx Series Dual Frequency Ultrasonic Cleaners,
  • With two holes
Specifications
Product Type Lid
Capacity (Metric) 3.78 L
Capacity (English) 1 gal.
For Use With (Equipment) FB11211 Ultrasonic Cleaning Unit
Material Stainless Steel