Learn More
Fisherbrand™ 384-Well Polystyrene Plates
Product Code. 13565440 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Clear
White
Well Shape:
Rounded Square
Triangular, Flat Bottom (F-well)
Unit Size:
Pack of 100
Pack of 90
Product Code. Color Well Shape unitSize
13565440 Clear Triangular, Flat Bottom (F-well) Pack of 100
11947954 Clear Rounded Square Pack of 90
13545440 White Rounded Square Pack of 100
13555440 Black Rounded Square Pack of 100
4 options
Product Code. 13565440
Only null left
Add to basket
Add to basket
Process four times as many samples as standard 96-well plates
Optimized for robotic instruments, plate readers and liquid handling systems.
  • Highly specified polystyrene
  • Ideal for low-volume assays such as colorimetric, enzymatic, cloning, replication, and array, as well as storage
  • Flat well bottom surface provides greater area for binding
  • Working volume: 10–100μL
  • 384-well microplates conserve samples and reagents, providing cost savings and greater screening productivity
  • Untreated
  • Non-sterile
Order Info
Accommodate 12-565-511 lid (sold separately).
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Array 16 x 24
Color Clear
Product Type 384-Well Microplate
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 10 to 100 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Triangular, Flat Bottom (F-well)
For Use With (Application) For low volume assays such as colorimetric, enzymatic, cloning, replication and array, as well as storage
Quantity 180/Cs.