Learn More
Fisherbrand™ Traceable™ Digital Thermometers with Short Sensors
Product Code. 11799735 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Temperature Range (Metric):
-5°C to +50°C
-50°C to +70°C
Temperature Range (English):
-58°F to +158°F
23°F to 122°F
N/A
Scale:
Celsius
Celsius, Fahrenheit
Length (Metric):
57 mm
70 mm
102 mm
108 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Temperature Range (English) Scale Length (Metric) Unit Size
11799735 -50°C to +70°C -58°F to +158°F Celsius, Fahrenheit 70 mm Each
11789755 -50°C to +70°C -58°F to +158°F Celsius, Fahrenheit 102 mm Each
11709765 -50°C to +70°C N/A Celsius 102 mm Each
11759755 -5°C to +50°C 23°F to 122°F Celsius, Fahrenheit 108 mm Each
11735843 -50°C to +70°C -58°F to +158°F Celsius, Fahrenheit 57 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11799735
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
Fisherbrand™ Traceable™ Digital Thermometers are ideal for monitoring liquids, air/gas or semisolids.
Choose from models with a variety of features including MAX/MIN function to record the maximum and minimum temperature in a certain time period; visual and audible alarms and multiple displays of recorded temperatures.
  • Select the correct temperature to use in water baths, refrigerators, incubators and tanks
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™ indicates traceability to standards provided by NIST™.
Specifications
Certifications/Compliance ISO 17025, A2LA, NIST
Diameter (English) Probe 0.31 in.
Dimensions (L x W x H) 7 x 10.8 x 1.9 cm (2.75W x 4.25L x 0.75 in. D)
For Use With (Application) Monitoring liquids, air/gas or semisolids in water baths, refrigerators, freezers, incubators and tanks
Includes Sensor, AAA alkaline battery, flip open stand, wall mount, magnet and hook and loop mounting tape
Length (English) Cable 10 ft.
Length (English) Probe 0.75 in.
Length (Metric) 70 mm
Length (Metric) Cable 305 cm
Length (Metric) Probe 1.9 cm
Material Plastic
Resolution 1°C
Weight (English) 4 oz.
Weight (Metric) 113.3 g
Max. Temperature (English) 158°F
Max. Temperature (Metric) 70°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +158°F
Temperature Range (Metric) -50°C to +70°C
Calibrated Yes
Accuracy ±1°C
Display Digital LCD
Width (Metric) 15 cm
Scale Celsius, Fahrenheit