Learn More
Memmert™ Extension of DAkkS Factory Calibration for IF110
Product Code. 17526625 Shop All Memmert Products
Click to view available options
Unit Size:
Each
Product Code. 17526625
Brand: Memmert™ IF110E39697
Extension of DAkkS factory calibration by an additional freely selectable temperature for Memmert Incubators.
  • Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
For Use With (Equipment) IF110 Incubator
Product Type Factory Calibration Extension