Learn More
Wheaton™ Extended Controlled Dropper Tip
Shop All Wheaton Science Products Products
Click to view available options
Diameter (Metric):
13 mm (Tip)
20 mm (Tip)
8 mm (Tip)
Color:
Natural
Purple
White
Yellow
Unit Size:
Pack of 100
Specifications
Specifications
| Color | Natural |
| Material | LDPE |
| Drop Size | 41 μL |
| Product Type | Extended Controlled Dropper Tip |
| Diameter (Metric) | 13 mm (Tip) |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Documents
Scientific Resources
Catalogs
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
WHE026 Catalogue 2012 FINAL LOWRES
File size: 14.8 MB
Product Content Correction
Product Title
Wheaton™ Extended Controlled Dropper Tip >