Learn More
Eutech™ KCl Calibration Solution
Click to view available options
Quantity:
60 mL
480 mL
Conductivity:
100 ÎĽS/cm
111.8 mS/cm
12.88 mS/cm
1413 ÎĽS/cm
2764 ÎĽS/cm
500 ÎĽS/cm
84 ÎĽS/cm
Unit Size:
100mL
480mL
500mL
60mL
Description
Eutech™ KCl Calibration Solutions are used for calibrating conductivity probes for accurate measurement. Get quick and accurate conductivity measurements using Eutech™ conductivity meters and probes in the field, laboratories, and schools.
Features:
- Packed in 60mL or 480 ml durable plastic bottles
- Economical and convenient to use
Specifications
Specifications
Description | 111.8 mS/cm KCl calibration solution |
Conductivity | 111.8 mS/cm |
pH | 7 |
Boiling Point | 102.4°C |
For Use With (Equipment) | Conductivity Meters and Electrodes |
Solubility | Soluble |
Solution Type | Calibration Solution |
Type | Conductivity Standard |
Industry Type | Economical and Convenient for Laboratory Use |
Quantity | 480 mL |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
514-01 | Certificate of Analysis | 17/12/2024 | 01X211244 |
414-01 | Certificate of Analysis | 14/10/2024 | 01X211244 |
214-01 | Certificate of Analysis | 21/05/2024 | 01X211244 |
214-02 | Certificate of Analysis | 21/05/2024 | 01X211244 |
174-03 | Certificate of Analysis | 29/04/2024 | 01X211244 |
Product Content Correction
Product Title
Eutech™ KCl Calibration Solution >