Learn More
Tocris Bioscience™ E 64d
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Inhibitor of cathepsins B and L; also thought to inhibit calpain. Inhibits lysosomal proteases and interferes with autolysosomal digestion when used in combination with pepstatin A (Cat. No. 1190). Lysosome and cell permeable.
Specifications
Specifications
| CAS | 88321-09-9 |
| Quantity | 1 mg |
| Molecular Formula | C17H30N2O5 |
| Formula Weight | 342.43 |
| Purity | >98% |
| Molecular Weight (g/mol) | 342.436 |
| Chemical Name or Material | E 64d |