missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DY 268

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
DY 268 is a potent FXR antagonist (IC50 = 7.5 nM). Exhibits no detectable cytotoxicity in cell-based assays.
Specifications
Specifications
| Chemical Name or Material | 1-[(3-Methoxyphenyl)methyl]-N-[4-methyl-3-(4-morpholinylsulfonyl)phenyl]-3-(4-methylphenyl)-1H-pyrazole-4-carboxamide |
| CAS | 1609564-75-1 |
| Quantity | 50 mg |
| Target | LXR-like Receptor Antagonists |
| Molecular Formula | C30H32N4O5S |
| Formula Weight | 560.66 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction