Learn More
Fisherbrand™ Digital Thermometers with Stainless-Steel Probe on Cable
Product Code. 11779725 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Temperature Range (Metric):
-50°C to +150°C
-50°C to +260°C
-50°C to +300°C
0°C to 200°C
Temperature Range (English):
-58°F to +302°F
-58°F to +500.8°F
-58°F to +572°F
32°F to 392°F
Accuracy:
±1°C
±1°C between -30 to 150°C, otherwise ±2°C
±1°C from -20°C to +100°C
±2°C
Length (Metric) Cable:
45.72 cm
73.7 cm
106.6 cm
304.8 cm
305 cm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Temperature Range (English) Accuracy Length (Metric) Cable Unit Size
11779725 -50°C to +300°C -58°F to +572°F ±1°C from -20°C to +100°C 73.7 cm Each
15243986 -50°C to +260°C -58°F to +500.8°F ±1°C 304.8 cm Each
11715843 -50°C to +300°C -58°F to +572°F ±1°C 305 cm Each
11749725 -50°C to +150°C -58°F to +302°F ±1°C between -30 to 150°C, otherwise ±2°C 45.72 cm Each
11729735 0°C to 200°C 32°F to 392°F ±2°C 106.6 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11779725
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket
Easy-to-read digital thermometers are compact to hold in your hand or mount on wall in restricted areas
Easy-to-read digital thermometers are compact to hold in your hand or mount on wall in restricted areas. Fisher Scientific™ Digital thermometer with stainless-steel probe is chemically resistant to most acids, bases and solvents and may be used to measure temperature of gases, liquids, food, and other semisolids.
  • Models 15-077-16, -15, -21, -8B, -29 and 14-648-46 have alarm that can be set to alert user to temperatures that exceed setpoints
  • Some models (such as 15-077-16 and -15) have special features that print temperature, high/low memory and time of day; 15-077-29 thermometer has additional timer alarms
  • For a miniature-size thermometer, see model 15-077-32
For use in cuvets, soil, incubators, petroleum products and piercing food samples. Waterproof thermometers (models 15-077-9E, -9D and 14-648-46) are ideal for wet labs, wash-down areas and outdoor applications in the agricultural, petrochemical, food, and pharmaceutical industries.
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™; indicates traceability to standards provided by NIST™.
Specifications
Certifications/Compliance ISO 17025, A2LA, NIST
Diameter (English) Probe 0.19 in.
Dimensions (L x W x H) 4 x 3.25 x 4.75 in. (10 x 8.2 x 12.06 cm)
For Use With (Application) Wet labs, wash down areas and outdoor applications in the agricultural, petrochemical, food and pharmaceutical industries
Height (English) 4.75 in.
Height (Metric) 12.06 cm
Includes Traceable certificate, Stainless Steel probe, cable and Silver Oxide battery
Length (English) Cable 29 in.
Length (English) Probe 7.25 in.
Length (Metric) 100 mm
Length (Metric) Cable 73.7 cm
Length (Metric) Probe 18.41 cm
Material Plastic
Resolution 0.1°C
Weight (English) 4 oz.
Weight (Metric) 110 g
Width (English) 3.25 in.
Product Type Traceable Digital Thermometer w/Probe
Length (English) 4 in.
Max. Temperature (English) 572°F
Max. Temperature (Metric) 300°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Temperature Range (English) -58°F to +572°F
Temperature Range (Metric) -50°C to +300°C
Calibrated Yes
Accuracy ±1°C from -20°C to +100°C
Display Digital LCD
Width (Metric) 8.2 cm