Learn More
OHAUS™ Defender™ 5000 Washdown - D52 Low Profile Bench Scale, Models TD52XW QL Product Code.: 15918615
OHAUS™ Defender™ 5000 Washdown - D52 Low Profile Bench Scale, Models TD52XW QL
Product Code. 15918615 Shop All Ohaus Products
D52XW30WQL5-EU, 1 unit
Click to view available options
Model:
D52XW15WQDL5-EU
D52XW15WQDL5-M
D52XW15WQL5-EU
D52XW15WQL5-M
D52XW30WQDL5-M
D52XW30WQL5-EU
D52XW30WQL5-M
D52XW60WQDL5-EU
D52XW60WQDL5-M
D52XW60WQL5-EU
D52XW60WQL5-M
Unit Size:
1 unit
Product Code. 15918615
Brand: OHAUS™ 30471126
OHAUS™ Defender™ 5000 Series multifunctional bench scale is ideal for a multitude of applications. Built durably and equipped with advanced features such as maximum configurability print output, GMP/GLP data output, library/user management and multiple connectivity options.
  • User-friendly model with an easy-to-use keypad, large backlit display and plain text on-screen prompts which guide you through operation.
  • Durable construction with a stainless steel platform, frame, IP67 load cell and IP68 indicator.
  • Defender meets NTEP/Measurement Canada or OIML/EC Type approvals (depending on region).
  • Features common industrial modes and supports a 50,000-item library with 1,000 user profiles for easy data management.
  • Incudes GMP/GLP with time/date for result traceability.
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 20 Hours with Rechargeable Battery
Connection Type Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock
Construction 304-grade stainless steel platform and frame, stainless steel IP68 indicator, IP67 load cell, non-slip adjustable rubber feet
Capacity 30.00 kg
Display Type Backlit LCD
Readability 2.000000 g
Model D52XW30WQL5-EU
Protection Class Stainless Seel IP68 Indicator, IP67 Load Cell
Temperature (Metric) Operating -10°C to +40°C
Weighing Modes Custom, Gram, Kilogram, Metric Tonnes, Ounce, Pound, Pound:Ounce, Tonne
Dimensions (L x W x H) 677 x 400 x 122 mm
Weight (Metric) 17.240 kg
Show More Show Less
  • Multiple Manufacturers
    Don't miss our end-of-year sale: save up to 50% on premium lab equipment and instruments.
    Promo Code: 24121
    Expires2025-12-20
Product Content Correction
Product Title
OHAUS™ Defender™ 5000 Washdown - D52 Low Profile Bench Scale, Models TD52XW QL >