Learn More
Cole-Parmer™ CPC™ NSH Non-spill Coupling Insert with Hose Barb
Non-spill design virtually eliminates spills, minimizes downtime and enhances operator safety
Brand:  Cole-Parmer™ 31603-68
Product Code. 15497363
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • NS6 Series couplings feature non-spill valves at a great price.
  • Use the NS6 when even a few drops of spillage pose problems regarding safety, media cost or environmental regulations.
  • Soft touch overmold makes these couplings comfortable in the hand and very attractive.
  • Lightweight, chemically resistant and easy to use.
  • Non-spill design.
  • Disconnect under pressure with no spills.
  • Color coding Instant visual differentiation of media lines.
  • Glass-filled polypropylene.
  • Durable and compatible with many chemicals.
  • Colder thumb latch.
  • One-hand connection and disconnection.
Specifications
Specifications
Quick-Disconnect x Barb
1/2 in.
1/2 in. Quick Disconnect
Polypropylene
1/2 in.
Non-Spill, Quick-Disconnect x Compression Coupling Insert
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cole-Parmer™ CPC™ NSH Non-spill Coupling Insert with Hose Barb > 1/2 in.