missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CP 43
GREENER_CHOICE

Product Code. p-7145995
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18773459

Brand: Tocris Bioscience™ 6558/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent TAOK inhibitor

CP 43 is a potent TAO kinase (TAOK) inhibitor (IC50 values are 11 nM/L and 15 nM/L for TAOK1 and TAOK2, respectively). Inhibits TAOK phosphorylation and activation of JNK in vitro. Prolongs duration of mitosis and increases mitotic cell death in the SKBR3 breast cancer cell line. Also reduces tau phopshorylation in cortical neurons at sites associated with neurodegeneration in human tauopathies.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-Oxo-2-((1,2,3,4-tetrahydronaphthalen-1-yl)amino)ethyl)-[1,1′-biphenyl]-4-carboxamide
CAS 850467-66-2
Quantity 10 mg
Molecular Formula C25H24N2O2
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.