Learn More
Fisherbrand™ Cooling of Drain Water for Machines with High Speed Module and Condenser Recycle
Product Code. 17107854 Shop All Fisherbrand Products
Click to view available options
For Use With (Equipment):
Blue Line 5 / IQ5
Blue Line 6 / IQ6
Unit Size:
Each
Product Code. 17107854
Only null left
Add to basket
Add to basket
The drain water is mixed with cold water before entering the drain.
  • Water is cooled to ≤ 60°C
Specifications
For Use With (Application) Cooling Drain Water
For Use With (Equipment) Blue Line 6 / IQ6