Learn More
Fisherbrand™ Condenser Kit for Condenser Recycle
Product Code. 17197844 Shop All Fisherbrand Products
Click to view available options
:
For Condenser Recycle
Product Code. 17197844
Only null left
Add to basket
Add to basket
Standard condenser that collects water in a tank rather than letting the water out into the drain.
  • Collected water can then be used for washing in the next cycle
  • Tank includes a heating element, which keeps the temperature above 60°C and ensures that there is no possibility to develop bacteria in the tank
Specifications
Product Type Condenser Kit
For Use With (Equipment) Condenser Recycle