Learn More
Fisherbrand™ Combined Ion-Selective Electrode
Product Code. 11769708 Shop All Fisherbrand Products
Click to view available options
Electrode:
Ammonium Combined ISE
Chlorine Combined ISE
Copper Combined ISE
Nitrate Combined ISE
Unit Size:
Each
Product Code. 11769708
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Converts activity of a specific ion dissolved in a solution into an electrical potential. Fisherbrand™ Combined Ion-Selective Electrode is compatible with all pH/mV meters or ionometers.
  • With solid membrane Interchangeable flat surface
  • Ion to be detected passes through the membrane generating a difference of potential, proportional to ionic concentration
  • Analysis time is generally less than one minute
  • Sample preparation steps such as filtering or Distillation rarely necessary
  • Calibration should be carried out as often as possible (once a day) at the same temperature as the sample
  • Measurement temperature 0 to 50°C
  • Delivered with membrane, electrolytic solution, ISAB solution and ISE standard and gasket
ISAB solution:
To adjust the ionic strength. add directly to the solution to be measured.
ISE standard:
For the calibration of ionic electrodes. Concentration 1000 ppm.
Specifications
Applications Determination of Chlorine (Cl-) ion
Filling Solution Type ISAB/ISE
pH Range 2 to 12
Interferences S2-, I-, CN-, Br-
Electrode Chlorine Combined ISE
Parameter Chlorine Ion