Learn More
BRAND™ Class A Borosilicate Volumetric Flask with Polypropylene Stopper
Borosilicate volumetric flask with polypropylene stopper and USP single batch certificate.
Brand: BRAND™ 236953
700.87 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Borosilicate 3.3
- To DIN EN ISO 1042
- Calibrated to contain (TC, In)
- Includes USP certificate single batch delivery
Specifications
1 L | |
Autoclavable | |
Yes | |
Non-sterile |
Borosilicate Glass (3.3) | |
Reusable | |
Narrow | |
Flat |
Product Content Correction
Product Title
BRAND™ Class A Borosilicate Volumetric Flask with Polypropylene Stopper > Volumetric Flask