Learn More
OHAUS™ Centrifuge Bucket Accessory
Accessory for centrifuges
5990.00 NOK - 12895.00 NOK
Specifications
For Use With (Equipment) | Centrifuge Multi FC5916R 120 V and Centrifuge Multi FC5916 120 V |
---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
16692754
|
OHAUS™
30553104 |
MTP x2 V1 |
12895.00 NOK
Pack of 2 |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16632764
|
OHAUS™
30553119 |
MTP x2 V2 |
5990.00 NOK
Pack of 2 |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16642764
|
OHAUS™
30553121 |
FB x2 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Accessory for centrifuges
- Centrifuge Multi FC5916R 120V
- Centrifuge Multi FC5916 120V
Specifications
Centrifuge Multi FC5916R 120 V and Centrifuge Multi FC5916 120 V |