Learn More
OHAUS™ Centrifuge Bucket Accessory
Accessory for centrifuges
Brand: OHAUS™ 30553121
Additional Details : Weight : 1.98000kg
Description
- Accessory for centrifuges
- Centrifuge Multi FC5916R 120V
- Centrifuge Multi FC5916 120V
Specifications
Centrifuge Multi FC5916R 120 V and Centrifuge Multi FC5916 120 V | |
750 mL | |
99 mm | |
FB x2 |