Learn More
Invitrogen™ CellVue™ Jade Cell Labeling Kit
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
Description
- The Mini CellVue™ Kits are supplied with one vial of dye stock (1 mM in ethanol) and one vial of labeling vehicle (Diluent C)
- CellVue™ Jade is optimally excited at 478nm and has a peak emission of 508nm
- Compatible with most multi-color applications; however, due to similar emission properties it cannot be used in combination with FITC or Alexa Fluor™ 488 reagents
Specifications
Specifications
Quantity | 1 kit |
Reagent Type | Cell Tracker Compounds, Cell Labeling Reagents |
Product Type | Cell Labeling |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Invitrogen™ CellVue™ Jade Cell Labeling Kit >