missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Celastrol
GREENER_CHOICE

Product Code. p-4783839
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18712564

Brand: Tocris Bioscience™ 3203/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits TNF-α-induced NF-κB activation

Antioxidant and anti-inflammatory agent. Potently inhibits lipid peroxidation in mitochondria and inhibits TNF-α-induced NFκB activation. Also shown to inhibit topoisomerase II activity in vitro (IC50 = 7.41 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34157-83-0
Molecular Formula C29H38O4

Specifications

Quantity 10 mg
Formula Weight 450.61
Purity >99%
Molecular Weight (g/mol) 450.62
Chemical Name or Material Celastrol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.