missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CDK8/19i

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CDK8/19i is a potent and selective CDK8 and CDK19 inhibitor (IC50 values are 2.9 and 14.1 nM, respectively). CDK8/19i maintains pluripotency of mouse PSCs in culture. It induces the formation of dome-shaped colonies and upregulates Nanog-GFP reporter and endogenous Nanog expression in mouse ESCs in vitro.
Specifications
Specifications
| Chemical Name or Material | N-Propyl-3-(4-pyridinyl)imidazo[1,2-b]pyridazin-6-amine |
| CAS | 923596-52-5 |
| Quantity | 10 mg |
| Target | Cyclin-dependent Kinase Inhibitors |
| Molecular Formula | C14H15N5 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction