Learn More
Tocris Bioscience™ CB 65
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CB 65 is a high affinity and selective CB2 receptor agonist; Ki values are 3.3 and > 1000 nM for CB2 and CB1 receptors respectively.
Specifications
Specifications
| Chemical Name or Material | N-Cyclohexyl-7-chloro-1-[2-(4-morpholinyl)ethyl]quinolin-4(1H)-one-3-carboxamide |
| CAS | 913534-05-1 |
| Quantity | 10 mg |
| Target | Cannabinoid R2/CB2 Agonists |
| Molecular Formula | C22H28ClN3O3 |
| Purity | 0.99 |