Learn More
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed Product Code.: 10365511
Estimated Shipment: 21-10-2025
  • ONLINE EXCLUSIVE PRICE
    1675.00 NOK
    1626.13 NOK / Pack of 1000
    Save 48.87 NOK
    2% Off
Only null left
Add to basket
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Product Code. 10365511
150 mm, 1 mm, Pack of 1000
Click to view available options
Length (Metric):
10 mm
70 mm
75 mm
80 mm
90 mm
100 mm
120 mm
150 mm
Diameter (Metric) Outer:
1 mm
1.35 mm
1.55 mm
1.75 mm
2.35 mm
Unit Size:
Pack of 1000
Product Code. 10365511
Brand: Hirschmann™ 9211015
  • ONLINE EXCLUSIVE PRICE
    1675.00 NOK
    1626.13 NOK / Pack of 1000
    Save 48.87 NOK
    2% Off
Only null left
Estimated Shipment: 21-10-2025
Add to basket
Only null left
Add to basket
Excellent for melting point analysis in the laboratory. Hirschmann Instruments™ Capillary Tubes for Melting Point Determination, One End Closed are manufactured from clear glass.
  • Capillary tubes
  • Available in a range of diameters/lengths
Specifications
Diameter (Metric) Inner 0.8 mm
Diameter (Metric) Outer 1 mm
For Use With (Application) Melting point determination
Tube Style One end closed
Length (Metric) 150 mm
Type Capillary Tube
Product Content Correction
Product Title
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed >