missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calnexin Antibody (S07-2B1), Novus Biologicals™
Rabbit Monoclonal Antibody
3895.00 NOK - 5975.00 NOK
Specifications
| Antigen | Calnexin |
|---|---|
| Clone | S07-2B1 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18634105
|
Novus Biologicals
NBP3-19587-50ul |
50 μg |
3895.00 NOK
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681035
|
Novus Biologicals
NBP3-19587-100ul |
100 μg |
5975.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calnexin Monoclonal antibody specifically detects Calnexin in Human, Rat samples. It is validated for Western BlotSpecifications
| Calnexin | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
| A synthetic peptide of human Calnexin (Uniprot # P27824) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S07-2B1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 821 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title