missing translation for 'onlineSavingsMsg'
Learn More

Calnexin Antibody (S07-2B1), Novus Biologicals™

Product Code. 18681035 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
Product Code. Quantity unitSize
18681035 100 μg 100µL
18634105 50 μg 50µL
2 options
This item is not returnable. View return policy

Product Code. 18681035

Brand: Novus Biologicals NBP319587100ul

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Monoclonal Antibody

Calnexin Monoclonal antibody specifically detects Calnexin in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Calnexin
Applications Western Blot
Classification Monoclonal
Clone S07-2B1
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Gene Alias calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Host Species Rabbit
Immunogen A synthetic peptide of human Calnexin (Uniprot # P27824)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision
Primary or Secondary Primary
Gene ID (Entrez) 821
Target Species Human, Rat
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.