Learn More
Calnexin Antibody (S07-2B1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19587-50ul
This item is not returnable.
View return policy
Description
Calnexin Monoclonal antibody specifically detects Calnexin in Human, Rat samples. It is validated for Western Blot
Specifications
| Calnexin | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| S07-2B1 | |
| Unconjugated | |
| calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
| A synthetic peptide of human Calnexin (Uniprot # P27824) | |
| 50 μg | |
| ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision | |
| 821 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |