Learn More
Calnexin Antibody (S07-2B1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19587-50ul
 View more versions of this product
Product Code. 18634105
  • 3895.00 NOK / 50µL
Description
Calnexin Monoclonal antibody specifically detects Calnexin in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calnexin
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Rat
Purified
Western Blot
S07-2B1
Unconjugated
calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
A synthetic peptide of human Calnexin (Uniprot # P27824)
50 μg
ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision
821
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.