Learn More
Buffer Solution pH3.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™ Product Code.: 10104823
Buffer Solution pH3.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Product Code. p-7073112 Shop All Fisher Chemical Products
500 mL, HDPE plastic bottle, 500mL
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
HDPE plastic bottle
Plastic container
Unit Size:
1 L
2.5 L
500mL
Product Code. 10104823
Brand: Fisher Chemical J/2810/08
Science Is Hard Enough Your Bioreagents Shouldn’t Be
Use promo code 24128 at checkout
View Available Promotions
Request Bulk or Custom Format
Chemical Identifiers
MDL Number 217672
Specifications
Color Colorless
pH 3
Physical Form Liquid
Quantity 500 mL
Packaging HDPE plastic bottle
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Product Code
Lot Number2545012 Certificate TypeCertificate of Analysis Date17/07/2025 Product CodeJ/2810/08,J/2810/17,J/2810/15
Lot Number2539420 Certificate TypeCertificate of Analysis Date09/06/2025 Product CodeJ/2810/08,J/2810/17,J/2810/15
  • Multiple Manufacturers
    Save up to 59% on some of our most popular products
    Promo Code: 24140
    Expires2025-08-24
  • Multiple Manufacturers
    Reliable performance. Special prices.Happier researchers.
    Promo Code24128
    Expires2025-08-31
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24157
    Expires2025-08-31
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/07/2025 and 31/08/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Buffer Solution pH3.00 (Phthalate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™ >