Learn More
Buffer Solution pH 4.01, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
For Use With (Application) | pH calibration |
pH | 4.01 |
Quantity | 500 mL |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Product Content Correction
Product Title
Buffer Solution pH 4.01, Reagecon™ >