missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BTZO 1
GREENER_CHOICE

Product Code. 18043862
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18043862

Brand: Tocris Bioscience™ 4204/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MIF binder

Macrophage migration inhibitory factor (MIF) binder (Kd = 68.6 nM). Activates antioxidant response element (ARE)-mediated gene expression. Suppresses oxidative stress-induced cardiomyocyte apoptosis in vitro. Cardioprotective.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99420-15-2
Molecular Formula C13H8N2OS

Specifications

Quantity 50 mg
Formula Weight 240.28
Purity >99%
Molecular Weight (g/mol) 240.28
Chemical Name or Material BTZO 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.