missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BPTU
GREENER_CHOICE

Product Code. p-7114662
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18768629

Brand: Tocris Bioscience™ 6078/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

P2Y1 allosteric antagonist

BPTU is an allosteric antagonist of P2Y1 (EC50 = 0.06-0.3 μM). Non-nucleotide ligand. Binds receptor outside of the helical bundle. Blocks inhibition of spontaneous contraction of rat and mouse colon induced by electrical field stimulation, nicotine and P2Y agonists. Antithrombotic; reduces platelet aggregation.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[2-[2-(1,1-Dimethylethyl)phenoxy]-3-pyridinyl]-N′-[4-(trifluoromethoxy)phenyl]urea
CAS 870544-59-5
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C23H22F3N3O3
Formula Weight Observed MW: 445.43
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.