Learn More
BRAND™ BLAUBRAND™ Amber Borosilicate Glass Class A Volumetric Flask with Glass Stopper Product Code.: 11844180
Only undefined left
Add to basket
BRAND™ BLAUBRAND™ Amber Borosilicate Glass Class A Volumetric Flask with Glass Stopper
Product Code. 11844180 Shop All Brand Products
250 mL, 14/23, Narrow, ±0.15 mL, Pack of 2
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
Closure Size:
10/19
12/21
14/23
19/26
24/29
Neck Style:
Narrow
Wide
Limit of Error:
±0.04 mL
±0.06 mL
±0.10 mL
±0.15 mL
±0.25 mL
±0.4 mL
Unit Size:
Each
Pack of 2
Product Code. 11844180
Brand: BRAND™ 37451
Only undefined left
Add to basket
Only undefined left
Add to basket
Manufactured from Amber Borosilicate Glass and compllies with DIN EN ISO 1042 standards. BRAND™ Blaubrand™ Amber Borosilicate Glass Class A Volumetric Flask with Glass Stopper protects light-sensitive liquids from deterioration.Batch number and certificate enclosed with packing. DakkS calibration certificate available upon request.
  • Manufactured from Amber Borosilicate Glass
  • Complies with DIN EN ISO 1042 standards.
  • Calibrated to contain (TC, In.)
  • Includes amber glass stopper, interchangeable with polypropylene stopper.
  • Dark amber flask protects light-sensitive liquids from deterioration.
  • Batch number and certificate enclosed with packing.
  • DakkS calibration certificate available upon request.
TRUSTED_SUSTAINABILITY
Specifications
Calibration Calibrated To Contain
Certifications/Compliance DIN EN ISO 1042. DAkkS calibration certificate upon request.
Closure Material Glass
Color Amber
Capacity (Metric) 250 mL
Material Borosilicate Glass
Limit of Error ±0.15 mL
Neck Style Narrow
No. per Case 2 Piece
Tolerance ±0.15 mL
Class A
Closure Size 14/23
Show More Show Less
  • Brand - BLAUBRAND
    BLAUBRAND video
    Video Player is loading.
    Current Time 0:00
    Duration 2:09
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 2:09
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Brand BLAUBRAND
Product Content Correction
Product Title
BRAND™ BLAUBRAND™ Amber Borosilicate Glass Class A Volumetric Flask with Glass Stopper >